1,10-Phenanthroline monohydrate

Supplier: Molekula
Danger

Synonyms: 1,10-Phenanthroline hydrate

MOLE15746698-25GEA 140 GBP
MOLE15746698-25G
1,10-Phenanthroline monohydrate
1,10-Phenanthroline monohydrate
Formula: C₁₂H₈N₂·1H₂O
MW: 198,22 g/mol
Melting Pt: 93…94 °C
Density: 1,259 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149973
CAS Number: 5144-89-8
EINECS: 200-629-2
UN: 2811
ADR: 6.1,III
Merck Index: 12,07356
https://ni.vwr.com/stibo/web/std.lang.all/19/91/2351991.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR