3,3',5,5'-Tetramethylbenzidine (TMB) substrate

Supplier: ENZO LIFE SCIENCES AG

Synonyms: TMB3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine

ENZOADI800350EA 55.1 GBP
ENZOADI800350 ALEXADI-80-0350
3,3',5,5'-Tetramethylbenzidine (TMB) substrate
3,3',5,5'-Tetramethylbenzidine (TMB)

HRP substrate.


  • Used in ELISA, colorimetric detection
  • Solution of 3,3',5,5'-tetramethylbenzidine (TMB) and hydrogen peroxide

Formula: C₁₆H₂₀N₂
MW: 240,35 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00007748
CAS Number: 54827-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR