D-(+)-Malic acid, Supelco®

Supplier: Merck
Warning

Synonyms: (R)-(+)-Hydroxysuccinic acid, D(+)-Hydroxy-1,4-butanedioic acid, (R)-(+)-Hydroxy-1,4-butanedioic acid, D(+)-Malic acid, D(+)-Hydroxysuccinic acid, (R)-2-Hydroxysuccinic acid

M-Clarity™ quality level = MQ100

46940-U
46940-UEA 43.4 GBP
46940-U
D-(+)-Malic acid, Supelco®
D(+)-Malic acid
Formula: C₄H₆O₅
MW: 134,09 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 101…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00004245
CAS Number: 636-61-3
EINECS: 211-262-2
Merck Index: 13,05307
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e8e8543edbc1ac226f6b4b119c1606144c206db&vid=4e8e8543edbc1ac226f6b4b119c1606144c206db&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR