Talc, powder, Multi-Compendial, J.T.Baker®
Supplier: Avantor
|
Warning
|
Synonyms:
Soapstone
, Magnesium silicate monohydrate
, Talcum venetum
4100-REA
1520
GBP
4100-R
4100-05
4100-01
Talc, powder, Multi-Compendial, J.T.Baker®
Talc
Formula:
Mg₃O₁₁Si₄·1H₂O MW: 379,27 g/mol Melting Pt: 1200 °C Density: 2,58…2.83 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00792903 CAS Number: 14807-96-6 EINECS: 238-877-9 Merck Index: 13,09127 |
Specification Test Results
GMP Manufactured Product | |
Meets U.S.P Requirements | |
Meets B.P. Chemical Specifications | |
Meets E.P. Chemical Specifications | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Identification C | Passes Test |
Microbial Limit - Total Aerobic Bacterial Count | ≤100 cfu/g |
Microbial Limit - Total Molds and Yeasts | ≤50 cfu/g |
USP - Acidity and Alkalinity | Passes Test |
USP - Loss on Ignition | ≤7.0 % |
USP - Water-Soluble Substances | ≤0.1 % |
USP - Iron (Fe) | ≤0.25 % |
USP - Lead (Pb) | ≤0.001 % |
USP - Calcium (Ca) | ≤0.9 % |
USP - Aluminum (Al) | ≤2.0 % |
USP - Absence of Asbestos (Method A) | Passes Test |
USP - Magnesium (as MgO) | 17.0 - 19.5 % |
EP/BP - Free from Asbestos | Passes Test |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Identification C | Passes Test |
EP/BP - Acidity or Alkalinity | Passes Test |
EP/BP - Water-Soluble Substances | ≤0.2 % |
EP/BP - Aluminum (Al) | ≤2.0 % |
EP/BP - Calcium (Ca) | ≤0.90 % |
EP/BP - Iron (Fe) | ≤0.25 % |
EP/BP - Lead (Pb) | ≤10.0 ppm |
EP/BP - Magnesium (Mg) | 17.0 - 19.5 % |
EP/BP - Loss on Ignition | ≤7.0 % |
EP/BP - Total Aerobic Microbial Count, cfu/g | ≤1000 |
EP/BP - Total Yeast and Mold Count, cfu/g | ≤100 |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. | |
Suitable for Oral Administration |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...