Talc
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Soapstone
, Magnesium silicate monohydrate
, Talcum venetum
40318.22EA
27.5
GBP
40318.22
40318.A4
40318.36
Talc
Talc
Lubricant, filler, electric and heat insulator, in clarifying liquids by filtration
Formula:
Mg₃O₁₁Si₄·1H₂O MW: 379,27 g/mol Melting Pt: > 1200 °C Density: 2,7 Storage Temperature: Ambient |
MDL Number:
MFCD00792903 CAS Number: 14807-96-6 EINECS: 238-877-9 Merck Index: 13,09127 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...