Hydrazinium sulphate 99+%

Supplier: Thermo Scientific
Danger

Synonyms: Hydrazine sulphate salt, Hydrazine sulphate

201861000 201865000
ACRO201861000EA 71.3 GBP
ACRO201861000 ACRO201865000
Hydrazinium sulphate 99+%
Hydrazinium sulphate
Formula: NH₂NH₂·H₂SO₄
MW: 130,12 g/mol
Melting Pt: 254 °C
Density: 1,37 g/cm³ (20 °C)
MDL Number: MFCD00044873
CAS Number: 10034-93-2
EINECS: 233-110-4
UN: 3288
ADR: 6.1,III
Merck Index: 14,04772
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e277940036b3cf71839ff7458f0c7d08bff6f6d5&vid=29f752de731326a8d6916b9e9efb1b4139b610a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration with NaOH ≥99.0 %
Total chloride ≤20 ppm
Calcium (Ca) ≤10 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤5 ppm
Magnesium (Mg) ≤5 ppm
Potassium (K) ≤10 ppm
Sodium (Na) ≤20 ppm
Zinc (Zn) ≤5 ppm
Total silicon (Si) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR