(±)-3-Quinuclidinol ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(±)-Quinuclidin-3-ol, (±)-3-Hydroxyquinuclidine
B21503.06
B21503.14
B21503.22
B21503.06EA
22.1
GBP
B21503.06
B21503.14
B21503.22
(±)-3-Quinuclidinol ≥98%
(±)-3-Quinuclidinol
Purity: 98+%
CAS No.: 1619-34-7
Molecular Formula: C7H13NO
Formula Weight: 127.19
pH: 10-12
Melting Point: 217-224°
Beilstein Reference: 104327
Merck Reference: 14,8082
UN No.: 3263
MDL No.: MFCD00151326
Formula:
C₇H₁₃NO MW: 127,19 g/mol Melting Pt: 217…224 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151326 CAS Number: 1619-34-7 EINECS: 216-578-4 UN: 3263 ADR: 8,III Merck Index: 14,8082 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...