(±)-Phenylsuccinic acid ≥98%
Supplier: Thermo Scientific
Synonyms:
(±)-Phenyl-1,4-butanedioic acid, Phenylsuccinic acid
A11971.14
A11971.22
A11971.36
A11971.14EA
25.7
GBP
A11971.14
A11971.22
A11971.36
(±)-Phenylsuccinic acid ≥98%
(±)-Phenylsuccinic acid
Purity: 98%
CAS No.: 635-51-8
Molecular Formula: C10H10O4
Formula Weight: 194.19
Melting Point: 167-169°
Beilstein Reference: 1875051
Formula:
C₁₀H₁₀O₄ MW: 194,19 g/mol Boiling Pt: 368 °C (1013 hPa) Melting Pt: 167…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004256 CAS Number: 635-51-8 EINECS: 211-238-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...