(±)-2-Tridecanol ≥97%

Supplier: Thermo Scientific

Synonyms: 2-Tridecanol, (±)-2-Hydroxy tridecane

L01926.06 L01926.03
L01926.06EA 72 GBP
L01926.06 L01926.03
(±)-2-Tridecanol ≥97%
(±)-2-Tridecanol

Purity: 97% CAS No.: 1653-31-2 Molecular Formula: C13H28O Formula Weight: 200.37 Refractive Index: 1.4430 Melting Point: 23-26° Boiling Point: 265° Beilstein Reference: 1720217 MDL No.: MFCD00037601

Formula: C₁₃H₂₈O
MW: 200,36 g/mol
Boiling Pt: 265 °C
Melting Pt: 23…26 °C
Density: 0,83
Storage Temperature: Ambient
MDL Number: MFCD00037601
CAS Number: 1653-31-2
EINECS: 216-723-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR