trans-Dichloroethylene 99+% stabilised
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(E)-1,2-Dichloroethene, trans-Dichloroethene, (E)-1,2-Dichloroethylene, trans-1,2-Dichloroethylene
406840250
406842500
ACRO406840250EA
139
GBP
ACRO406840250
ACRO406842500
trans-Dichloroethylene 99+% stabilised
trans-Dichloroethylene
Formula:
C₂H₂Cl₂ MW: 96,94 g/mol Boiling Pt: 48 °C (760 mmHg) Melting Pt: –50 °C Density: 1,26 g/cm³ Flash Pt: 6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00062942 CAS Number: 156-60-5 EINECS: 205-860-2 UN: 1150 ADR: 3,II Merck Index: 12,00093 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4450 to 1.4470 (20°C, 589 nm) |
GC | ≥99.0 % |
Stabilizer | 85 to 115 ppm MEHQ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...