(±)-Mercaptosuccinic acid ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: (±)-Mercapto-1,4-butanedioic acid, 2-Mercaptosuccinic acid, (±)-Thiomalic acid, DL-Mercaptosuccinic acid

B23301.14 B23301.22 B23301.36
B23301.14EA 26.2 GBP
B23301.14 B23301.22 B23301.36
(±)-Mercaptosuccinic acid ≥98%
(±)-Mercaptosuccinic acid

Purity: 98% CAS No.: 70-49-5 Molecular Formula: C4H6O4S Formula Weight: 150.15 Melting Point: 150-155° Beilstein Reference: 1099858 Merck Reference: 14,9343 MDL No.: MFCD00004860

Formula: C₄H₆O₄S
MW: 150,16 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 150…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00004860
CAS Number: 70-49-5
EINECS: 200-736-4
Merck Index: 14,9343

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR