(±)-Mercaptosuccinic acid 99%

Supplier: Thermo Scientific
Warning

Synonyms: (±)-Mercapto-1,4-butanedioic acid, 2-Mercaptosuccinic acid, (±)-Thiomalic acid, DL-Mercaptosuccinic acid

125551000 125555000 125550025
ACRO125551000EA 104 GBP
ACRO125551000 ACRO125555000 ACRO125550025
(±)-Mercaptosuccinic acid 99%
(±)-Mercaptosuccinic acid
Formula: C₄H₆O₄S
MW: 150,16 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 152…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00004860
CAS Number: 70-49-5
EINECS: 200-736-4
Merck Index: 12,09479

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 150 to 155 °C
Titration with NaOH 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR