Polyethylene glycol 12,000
Supplier: Thermo Fisher Scientific
Synonyms:
PEG 12,000
42635.30
42635.A1
42635.30EA
60.5
GBP
42635.30
42635.A1
Polyethylene glycol 12,000
Polyethylene glycol 12,000
CAS No.: 25322-68-3
Molecular Formula: H(OCH2CH2)nOH
Flash Point: ≥240°(464°F)
Physical Form: Powder
Odour: Odourless
pH: 5-7
Melting Point: 64-65°
Merck Reference: 14,7568
MDL No.: MFCD00081839
Formula:
H(OCH₂CH₂)nOH Melting Pt: 64…65 °C Flash Pt: >240 °C (464 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 500-038-2 Merck Index: 14,7568 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...