2'-Hydroxychalcone ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
L13937.06
L13937.14
L13937.06EA
12.3
GBP
L13937.06
L13937.14
2'-Hydroxychalcone ≥98%
2'-Hydroxychalcone
Purity: 98+%
CAS No.: 1214-47-7
Molecular Formula: C15H12O2
Formula Weight: 224.26
Melting Point: 89-91°
Beilstein Reference: 976324
Formula:
C₁₅H₁₂O₂ Melting Pt: 89…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016441 CAS Number: 1214-47-7 EINECS: 214-928-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...