2-Hydroxychalcone ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A12851.06
A12851.14
A12851.06EA
32.4
GBP
A12851.06
A12851.14
2-Hydroxychalcone ≥98%
2-Hydroxychalcone
Purity: 98+%
CAS No.: 644-78-0
Molecular Formula: C15H12O2
Formula Weight: 224.26
Melting Point: ca 154° dec.
Beilstein Reference: 2049134
Formula:
C₁₅H₁₂O₂ Melting Pt: ∼ 154 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00016449 CAS Number: 644-78-0 EINECS: 211-422-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...