Niobium ≥99.8% (metals basis), rods, annealed, Ø 6.35 mm (0.25 in)

Supplier: Thermo Fisher Scientific

10273.BQ 10273.BU 10273.BY
10273.BQEA 69.5 GBP
10273.BQ 10273.BU 10273.BY
Niobium ≥99.8% (metals basis), rods, annealed, Ø 6.35 mm (0.25 in)
Niobium

Purity: 99.8% CAS No.: 7440-03-1 Physical Form: ≈2.7g/cm, Temper: soft Odour: Odourless MDL No.: MFCD00011126

Formula: Nb
Boiling Pt: 3700 °C (1013 hPa)
Melting Pt: 2468 °C
Density: 8,57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR