Gadolinium ≥99.9% (metals basis excluding Ta), rods, Ø 19 mm (0.75 in)

Supplier: Thermo Scientific

43031.HF 43031.DH
43031.HFEA 317 GBP
43031.HF 43031.DH
Gadolinium ≥99.9% (metals basis excluding Ta), rods, Ø 19 mm (0.75 in)
Gadolinium

Purity: 99.9% CAS No.: 7440-54-2 Molecular Formula: Packaged in mylar under argon Physical Form: ≈2.2g/mm, Cast rod, diameter is approximate MDL No.: MFCD00011022

Formula: Gd
MW: 157,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011022
CAS Number: 7440-54-2
EINECS: 231-162-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR