Carbon ≥99%, felt, Thickness: 3.18 mm (0.125 in)
Supplier: Thermo Scientific
Synonyms:
Charcoal
43199.RY
43199.RR
43199.RYEA
61.1
GBP
43199.RY
43199.RR
Carbon ≥99%, felt, Thickness: 3.18 mm (0.125 in)
Carbon
Purity: 99.0%
Physical Form: ≈2.4g/10x10cm
MDL No.: MFCD00133992
Formula:
C MW: 12,01 g/mol Boiling Pt: ∼4000 °C (1013 hPa) Melting Pt: 3727 °C Density: 3,65 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133992 CAS Number: 7440-44-0 EINECS: 231-153-3 Merck Index: 13,01818 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...