Rivaroxaban 98%

Supplier: Thermo Fisher Scientific
Warning

461600010 461602500
ACRO461600010EA 495 GBP
ACRO461600010 ACRO461602500
Rivaroxaban 98%
Rivaroxaban
Formula: C₁₉H₁₈ClN₃O₅S
Melting Pt: 228…229 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11974010
CAS Number: 366789-02-8

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 228 to 232 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR