Artemether

Supplier: Thermo Fisher Scientific
Warning

460700050 460700250
ACRO460700050EA 66.4 GBP
ACRO460700050 ACRO460700250
Artemether
Artemether
Formula: C₁₆H₂₆O₅
MW: 298.38 g/mol
Melting Pt: 86…89 °C
Storage Temperature: Freezer
MDL Number: MFCD00866205
CAS Number: 71963-77-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 86 to 90 °C
Loss on drying ≤0.5 %
HPLC ≥97.0 %
Specific optical rotation +166° to +175° (20 °C, 589 nm) (1 % in ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR