EDTA magnesium dipotassium salt 97%

Supplier: Thermo Fisher Scientific
Warning

457941000 457945000
ACRO457941000EA 82.9 GBP
ACRO457941000 ACRO457945000
EDTA magnesium dipotassium salt 97%
EDTA magnesium dipotassium salt
Formula: C₁₀H₁₂K₂MgN₂O₈
Storage Temperature: Ambient
MDL Number: MFCD00054450
CAS Number: 15708-48-2

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Assay 96.0 to 104.0 %
Loss on drying 6.5 % to 10.5 %
pH 6.0 to 8.0 (5 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR