4-Nitro-o-toluic acid 98%

Supplier: Thermo Fisher Scientific
Warning

457810050 457810250 457811000
ACRO457810050EA 100 GBP
ACRO457810050 ACRO457810250 ACRO457811000
4-Nitro-o-toluic acid 98%
4-Nitro-o-toluic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: 151…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00210697
CAS Number: 1975-51-5

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR