Mirtazapine 98%
Supplier: Thermo Scientific
Synonyms:
1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepine
456170010
456170050
ACRO456170010EA
493
GBP
ACRO456170010
ACRO456170050
Mirtazapine 98%
Mirtazapine
Formula:
C₁₇H₁₉N₃ MW: 265,36 g/mol Melting Pt: 121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00865427 CAS Number: 85650-52-8 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 119 to 123 °C |
Water | ≤3.5 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...