Glycoluril 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Tetrahydroimidazo[4,5-d]imidazole-2,5(1H,3H)-dione, Acetylenediurea
120130250
120131000
120135000
ACRO120130250EA
47.9
GBP
ACRO120130250
ACRO120131000
ACRO120135000
Glycoluril 97%
Glycoluril
Formula:
C₄H₆N₄O₂ MW: 142,12 g/mol Boiling Pt: 862 °C (760 mmHg) Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005539 CAS Number: 496-46-8 |
Specification Test Results
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Assay | ≥96.0 % (N) |
NMR | consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...