Cold resistant gloves, uvex unilite thermo HD
Supplier: UVEX
6094208 6094211
UVEX6094208EA 0 GBP
UVEX6094208 UVEX6094211
Cold resistant gloves, uvex unilite thermo HD
Gloves
Despite its extremely robust coating, the uvex unilite thermo stays warm and flexible even in extreme cold. The two-layer material composition of acrylic and cotton terry ensures a pleasant hand climate and good wearer comfort at all times. The gloves feature a PVC coating on the palm and the back of the hand.
  • Extremely robust coating is flexible even at low temperatures
  • Double layer construction ensures effective insulation
  • Good grip in dry and slightly damp areas
  • Excellent dexterity, high flexibility
  • Mechanical protection for heavy-duty jobs
Ideal for forklift truck drivers and for use in refrigerated warehouses or when working in cold conditions. For use in damp and oily working conditions.
Certifications: EN 388:2016 (3231X), EN 511 (12X)