Nitrile gloves, Solo 977
Supplier: MAPA
34977006 34977007 34977008 34977009
MAPA34977006EA 31.2 GBP
MAPA34977006 MAPA34977007 MAPA34977008 MAPA34977009
Nitrile gloves, Solo 977
Gloves
These disposable nitrile gloves provide the best chemical protection in MAPA's disposable range.
  • Suitable for a wide range of chemicals
  • Good compromise between thickness, suppleness and resistance
  • Chlorinated interior for a easier doning
  • Pebbled on fingertips for a good grip
Typical applications include: Handling in laboratories, chemical industry, sampling chemicals, pharmaceutical preparation.
Certifications: EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUS; Cat. III