Sanitary heavy duty floor squeegee
Supplier: PERFEX CORP
9530 9518 9524
PERF9530EA 84.8 GBP
PERF9530 PERF9518 PERF9524
Sanitary heavy duty floor squeegee
Mops
These floor squeegees are non-sparking and chemical-resistant. Twin-mounted PVC blades act as a splash guard and are easily changed with the self-locking connection system.
  • Lightweight, durable blade and handle will not absorb bacteria, liquids, or odour
  • Unaffected by water, petroleum products, detergents, sanitisers, and solvents
  • Non-marring
  • Excellent tear resistance
  • Available in 18", 24" and 30" lengths
Ordering information: Polymer adjustable or fixed handle must be ordered separately.