Calcium oxide 95.0-100.5% FCC, Macron Fine Chemicals™

Supplier: Avantor Performance Materials

Synonyms: Quicklime, Calcium oxide from marble, caustic, Lime

4268-24EA 0 GBP
4268-24
Calcium oxide 95.0-100.5% FCC, Macron Fine Chemicals™
Calcium oxide
Formula: CaO
MW: 56,08 g/mol
MDL Number: MFCD00010911
CAS Number: 1305-78-8
UN: 1910
ADR: 8,III

Order Now

Specification Test Results

Food GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical 95.0 - 100.5 %
FCC - Assay (CaO) 95.0 - 100.5 %
FCC - Identification Passes Test
FCC - Acid-Insoluble Substances ≤1 %
FCC - Alkalies or Magnesium ≤3.6 %
FCC - Arsenic (As) ≤3 mg/kg
FCC - Fluoride (F) ≤0.015 %
FCC - Lead (Pb) ≤2 mg/kg
FCC - Loss on Ignition ≤10.0 %
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR