Vials, pathological, narrow neck
Supplier: AZPACK
215-2137 215-2138 215-2139
215-2137EA 668 GBP
215-2137 215-2138 215-2139
Vials, pathological, narrow neck
Bottles Glass Bottles
Type II soda-lime glass, clear.
  • Leakproof, maximum vial height of 83,5 mm and Ø of 29,5 mm
  • Available with or without aluminum screw caps
  • Only for use in applications without high thermal loads
Suitable for the collection, transport and storage of liquids and biological specimens.