Liquid nitrogen cylinders, low pressure, XL series
Supplier: TAYLOR WHARTON
L120-0C03TPED L120-0C01TPED L240-OCO1TPED L240-0C03TPED
478-0432EA 7510 GBP
478-0432 478-0431 478-0437 478-0436
Liquid nitrogen cylinders, low pressure, XL series
Dewars and Cryogenic Vessels
The Taylor Wharton XL series of low-pressure liquid gas cylinders are specifically designed for laboratory applications using LN₂. The XL-160, XL-180 and XL-240 allow for frequent usage, especially the pressure building models (which rapidly build pressure, thus reducing waiting time) and provide for larger on-site requirements. Ideal for use with the Taylor Wharton K-Series refrigerators.
  • Polished heavy duty outer body
  • Colour-coded relief valves for different pressure settings
  • Rugged transportable units
  • Castor-mounted for easy manoeuvrability (except XL-160)
Certifications: Approved according to Transportable Pressure Equipment Directive 1999/36 EC
Ordering information: Medium and high pressure vessels are also available (for liquid or gaseous nitrogen, argon or oxygen) - please ask for details. For the full range of accessories please contact your local Avantor office.