Neutral detergent, RBS T 230
Supplier: CHEMICAL
T230/E123 T230/E148
CHEPT230/E123EA 81 GBP
CHEPT230/E123 CHEPT230/E148
Neutral detergent, RBS T 230
Cleaning Agents Detergents and Surface Cleansers
For manual cleaning, soaking and ultrasonic cleaning of sensitive material, glassware and laboratory equipment.
  • Compatible with all materials
  • Concentrated solution
  • High emulsifying power
RBS T 230 is a neutral cleaning agent recommended for manual cleaning, soaking and ultrasonic cleaning of sensitive materials and laboratory equipment made of aluminum, zinc, and alloys that are sensitive to acids or bases. It efficiently removes organic and biological residues, distilation residues etc.
Ordering information: Available in box of 4 × 5 litres; 20 kgMore Product Information