Anti-Nephrocystin-5 Rabbit Polyclonal Antibody [clone: R467]

Supplier: Bioworld Technology
BS3319
BWRLBS3319EA 283 GBP
BWRLBS3319
Anti-Nephrocystin-5 Rabbit Polyclonal Antibody [clone: R467]
Antibodies
Synthetic peptide, corresponding to amino acids 432-486 of Human Nephrocystin-5.

Nephrocystin-5 (R467) pAb detects endogenous levels of Nephrocystin-5 protein.

Type: Primary
Antigen: Nephrocystin-5
Clonality: Polyclonal
Clone: R467
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR