SU-5402 ≥95%, Millipore®

Supplier: Merck Millipore Calbiochem

Synonyms: 3-[3-(2-Carboxyethyl)-4-methylpyrrol-2-methylidenyl]-2-indolinone, SU 5402, 3-(4-Methyl-2-((2-oxoindolin-3-ylidene)methyl)-1H-pyrrol-3-yl)propanoic acid

572630-1 572630-500
572630-1EA 788 GBP
572630-1 572630-500
SU-5402 ≥95%, Millipore®
SU-5402

SU5402 is a cell-permeable, reversible, and ATP-competitive inhibitor of the tyrosine kinase activity of Fibroblast growth factor receptor 1 (FGFR1). It also inhibits aFGF-induced tyrosine phosphorylation of ERK1 and ERK2.


  • Lyophilised solid
  • ≥95% by HPLC


This is only a weak inhibitor of tyrosine phosphorylation of the PDGF receptor and does not inhibit phosphorylation of the insulin receptor and the kinase activity of the EGF receptor.

MW: 296.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08235144
CAS Number: 215543-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR