Anti-HIF-PH2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-2219 NB100-2219SS
NOVUNB100-2219EA 715 GBP
NOVUNB100-2219 NOVUNB100-2219SS
Anti-HIF-PH2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal HIF Prolyl Hydroxylase 2 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunoprecipitation. Tested Reactivity: Mouse.

There is 1 scientific publication using this antibody.

Type: Primary
Antigen: HIF-PH2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR