Anti-ASC Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-78977 NBP1-78977SS
NOVUNBP1-78977EA 715 GBP
NOVUNBP1-78977 NOVUNBP1-78977SS
Anti-ASC Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal ASC/TMS1 Antibody. Tested Applications: Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (-). Tested Reactivity: Human, Mouse.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:400

Type: Primary
Antigen: ASC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR