Anti-RAD51D Mouse Monoclonal Antibody [clone: 5B3/6]

Supplier: Novus Biologicals
NB100-178 NB100-178SS
NOVUNB100-178EA 564 GBP
NOVUNB100-178 NOVUNB100-178SS
Anti-RAD51D Mouse Monoclonal Antibody [clone: 5B3/6]
Antibodies
Mouse Monoclonal Rad51D Antibody (5B3/6). Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human, Mouse (-).

Type: Primary
Antigen: RAD51D
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR