Anti-BAX Mouse Monoclonal Antibody (Biotin) [clone: BAX/962]

Supplier: Biotium
BNCB0962-500 BNCB0962-100
BNCB0962-500EA 538 GBP
BNCB0962-500 BNCB0962-100
Anti-BAX Mouse Monoclonal Antibody (Biotin) [clone: BAX/962]
Antibodies
Recognizes a protein of 21 kDa, identified as the Bax protein. This MAb is highly specific to Bax and shows no cross-reaction with Bcl-2 or Bcl-X protein. Bcl-2 blocks cell death following a variety of stimuli. Bax has extensive amino acid homology with Bcl-2 and it homodimerizes and forms heterodimers with Bcl-2. Overexpression of Bax accelerates apoptotic death induced by cytokine deprivation in an IL-3 dependent cell line, and Bax also counters the death repressor activity of Bcl-2.

Type: Primary
Antigen: BAX
Clonality: Monoclonal
Clone: BAX/962
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR