Anti-MYH11 Mouse Monoclonal Antibody (CF647) [clone: SMMS-1]

Supplier: Biotium
BNC470893-500 BNC470893-100
BNC470893-500EA 538 GBP
BNC470893-500 BNC470893-100
Anti-MYH11 Mouse Monoclonal Antibody (CF647) [clone: SMMS-1]
Antibodies
Smooth muscle myosin heavy chain (SM-MHC) is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmentally regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2. The antibody may be useful for the study of breast tumors as the presence of an intact layer of myoepithelial cells is an important feature, which may distinguish benign breast lesions and carcinoma in situ from invasive tumors.

Type: Primary
Antigen: MYH11
Clonality: Monoclonal
Clone: SMMS-1
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cat, Chicken, Cow, Dog, Guinea Pig, Human, Pig, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR