Anti-AR Mouse Monoclonal Antibody (CF594) [clone: DHTR/882]

Supplier: Biotium
BNC940882-500 BNC940882-100
BNC940882-500EA 538 GBP
BNC940882-500 BNC940882-100
Anti-AR Mouse Monoclonal Antibody (CF594) [clone: DHTR/882]
Antibodies
Recognizes a protein of 110 kDa, which is identified as androgen receptor (AR). It reacts with full length, and the newly described A form of the receptor. It does not cross react with estrogen, progesterone, or glucocorticoid receptors. The expression of AR is reportedly inversely correlated with histologic grade i.e. well differentiated prostate tumors show higher expression than the poorly differentiated tumors. In prostate cancer, AR has been proposed, as a marker of hormone-responsiveness and thus it may be useful in identifying patients likely to benefit from anti-androgen therapy. Anti-androgen receptor has been useful clinically in differentiating morpheaform basal cell carcinoma (mBCC) from desmoplastic trichoepithelioma (DTE) in the skin.This MAb is superb for staining of formalin/paraffin tissues.

Type: Primary
Antigen: AR
Clonality: Monoclonal
Clone: DHTR/882
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR