Anti-HSPD1 Mouse Monoclonal Antibody (CF594) [clone: LK1]

Supplier: Biotium
BNC940851-500 BNC940851-100
BNC940851-500EA 538 GBP
BNC940851-500 BNC940851-100
Anti-HSPD1 Mouse Monoclonal Antibody (CF594) [clone: LK1]
Antibodies
Recognizes a 60 kDa protein, identified as the heat shock protein 60 (hsp60). Its epitope is localized between aa 383-447 of human hsp60. A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of a family of proteins known as heat shock proteins (hsp), more appropriately called as stress response proteins (srp). hsp60 is a potential antigen in a number of autoimmune diseases. In human arthritis and in experimentally induced arthritis in animals, disease development coincides with the development of immune reactivity directed against not only bacterial hsp60, but also against its mammalian homolog. Clone LK1, unlike LK2, recognizes only the mammalian (not bacterial) hsp60 and is useful in distinguishing hsp60 from mammals and bacteria.

Type: Primary
Antigen: HSPD1
Clonality: Monoclonal
Clone: LK1
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chicken, Cow, Dog, Drosophila, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus laevis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR