Anti-VCAM1 Mouse Monoclonal Antibody (CF594) [clone: VCAM1/843]

Supplier: Biotium
BNC940843-500 BNC940843-100
BNC940843-500EA 538 GBP
BNC940843-500 BNC940843-100
Anti-VCAM1 Mouse Monoclonal Antibody (CF594) [clone: VCAM1/843]
Antibodies
Recognizes a protein of 110 kDa, identified as CD106 (also known as vascular cell adhesion molecule-1 (VCAM-1) and INCAM-100). CD106 is a member of the Ig superfamily of adhesion molecules and is expressed at high levels on cytokine stimulated vascular endothelial cells, and at minimal levels on un-stimulated endothelial cells. It is also present on follicular and inter-follicular dendritic cells of lymph nodes, myoblasts, and some macrophages. CD106 serves as a ligand for leukocyte integrin (VLA-4 or CD49d/CD29) and mediates cell adhesion of leukocytes to activated endothelium. It plays a role in various immunological and inflammatory responses.

Type: Primary
Antigen: VCAM1
Clonality: Monoclonal
Clone: VCAM1/843
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR