Anti-C2 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-8613R
BOSSBS-8613REA 648 GBP
BOSSBS-8613R
Anti-C2 Rabbit Polyclonal Antibody
Antibodies
Component C2 which is part of the classical pathway of the complement system is cleaved by activated factor C1 into two fragments: C2b and C2a. C2a, a serine protease, then combines with complement factor C4b to generate the C3 or C5 convertase.

Recommended Dilutions: Flow Cytometry: 1:20 to 100; IHC-P: 1:100 to 500; IF(IHC-P): 1:50 to 200.

Type: Primary
Antigen: C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR