Anti-KRT14 Mouse Monoclonal Antibody (Purified) [clone: KRT14/532]

Supplier: Biotium
BNUB0532-500 BNUB0532-100
BNUB0532-500EA 491 GBP
BNUB0532-500 BNUB0532-100
Anti-KRT14 Mouse Monoclonal Antibody (Purified) [clone: KRT14/532]
Antibodies
Cytokeratin 14 (CK14) belongs to the type I (or A or acidic) subfamily of low molecular weight keratins and exists in combination with keratin 5 (type II or B or basic). CK14 is found in basal cells of squamous epithelia, some glandular epithelia, myoepithelium, and mesothelial cells. Anti-CK14 is useful in differentiating squamous cell carcinomas from poorly differentiated epithelial tumors. Anti-CK14 is one of the specific basal markers for distinguishing between basal and non-basal subtypes of breast carcinomas. Anti-CK14 is also a good marker for differentiation of intraductal from invasive salivary duct carcinoma by the positive staining of basal cells surrounding the in-situ neoplasm as well as for differentiation of benign prostate from prostate carcinoma. Furthermore, this antibody has been useful in separating oncocytic tumors of the kidney from its renal mimics, and in identifying metaplastic carcinomas of the breast.

Type: Primary
Antigen: KRT14
Clonality: Monoclonal
Clone: KRT14/532
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR