Anti-INS Mouse Monoclonal Antibody (CF594) [clone: E2-E3]

Supplier: Biotium
BNC940281-500 BNC940281-100
BNC940281-500EA 538 GBP
BNC940281-500 BNC940281-100
Anti-INS Mouse Monoclonal Antibody (CF594) [clone: E2-E3]
Antibodies
Recognizes a polypeptide which is identified as insulin, a 51-amino acid polypeptide composed of A and B chains connected through the C-peptide. Proinsulin, which has very little biological activity, is cleaved by proteases within its cell of origin into the insulin molecule and the C-terminal basic residue. Insulin enhances membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides, and synthesis of proteins and nucleic acids. Deficiency of insulin results in diabetes mellitus. The main storage site for insulin is the pancreatic islets. Antibodies to insulin are important as beta-cell and insulinoma marker.

Type: Primary
Antigen: INS
Clonality: Monoclonal
Clone: E2-E3
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cow, Human, Mouse, Pig, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR