Anti-Testosterone Mouse Monoclonal Antibody (CF405S) [clone: 4E1G2]

Supplier: Biotium

BNC040250-500 BNC040250-100
BNC040250-500EA 510 GBP
BNC040250-500 BNC040250-100
Anti-Testosterone Mouse Monoclonal Antibody (CF405S) [clone: 4E1G2]
Antibodies
This MAb is highly specific to testosterone. Its affinity constant for testosterone is ~1010M-1. In competitive binding immunoassay, it reacts with testosterone 100%, 11-beta-hydroxy testosterone 3.3%, 17-alpha-methyl testosterone

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: 4E1G2
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: All species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR