Anti-HLA-DQ Mouse Monoclonal Antibody (CF594) [clone: SPV-L3]
Supplier: Biotium
BNC940200-500 BNC940200-100
BNC940200-500EA 538 GBP
BNC940200-500 BNC940200-100
Anti-HLA-DQ Mouse Monoclonal Antibody (CF594) [clone: SPV-L3]
Antibodies
Recognizes a DQ antigen, which is a dimer of 60 kDa. The class II molecule is a heterodimer consisting of an alpha (DQA) and a beta chain (DQB), both anchored in the membrane. It plays a central role in the immune system by presenting peptides derived from extracellular proteins. Class II molecules are expressed in antigen presenting cells (APC: B Lymphocytes, dendritic cells, macrophages). The alpha chain is approximately 33-35 kDa. It is encoded by 5 exons; exon 1 encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, and exon 4 encodes the transmembrane domain andMore Product Information