Anti-Lung Specific Antigen Mouse Monoclonal Antibody (CF594) [clone: LSA120]

Supplier: Biotium
BNC940120-500 BNC940120-100
BNC940120-500EA 538 GBP
BNC940120-500 BNC940120-100
Anti-Lung Specific Antigen Mouse Monoclonal Antibody (CF594) [clone: LSA120]
Antibodies
LSA120 MAb is directed against a lung cell antigen. It shows a well-defined reactivity with cells of the lung lineage at all stages of maturation. Non-lung lineages are negative by immuno-histology. Although the exact identity of its antigen has yet to be determined, it has been shown to be distinct from MOC-52 and UM-A9 proteins. This MAb is useful in the identification of normal and neoplastic human lung cells.

Type: Primary
Antigen: Lung Specific Antigen
Clonality: Monoclonal
Clone: LSA120
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR