Anti-Nucleolar / Nucleoli Mouse Monoclonal Antibody (CF594) [clone: NM95]

Supplier: Biotium
BNC940095-500 BNC940095-100
BNC940095-500EA 538 GBP
BNC940095-500 BNC940095-100
Anti-Nucleolar / Nucleoli Mouse Monoclonal Antibody (CF594) [clone: NM95]
Antibodies
This MAb is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells. This monoclonal antibody is part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. MAb NM95 recognizes an antigen associated with the nucleoli in human cells. It can be used to stain the nucleoli in cell or tissue preparations and can be used as a marker of the nucleoli in subcellular fractions. It produces a speckled pattern in the nuclei of cells of normal and malignant cells and may be used to stain the nucleoli of cells in fixed or frozen tissue sections. It can be used with paraformaldehyde fixed frozen tissue or cell preparations and formalin fixed, paraffin-embedded tissue sections.

Type: Primary
Antigen: Nucleolar / Nucleoli
Clonality: Monoclonal
Clone: NM95
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR