Anti-Granulocyte Marker Mouse Monoclonal Antibody (CF568) [clone: BM-1]

Supplier: Biotium
BNC680061-500 BNC680061-100
BTIUBNC680061-500EA 538 GBP
BTIUBNC680061-500 BTIUBNC680061-100
Anti-Granulocyte Marker Mouse Monoclonal Antibody (CF568) [clone: BM-1]
Antibodies
Recognizes 183 kDa protein with DNA-binding characteristics, which is identified as a myeloid specific antigen. It reacts with myeloid precursor cells and granulocytes in bone marrow. Its antigen appears to be restricted to M2 and M3 acute myelogenous leukemia (AML) subtypes. Markers of myeloid cells are useful in the identification of different levels of cellular differentiation. This MAb reacts with early precursor and mature forms of human myeloid cells. It is useful in the identification of myelogenous leukemias, distinguishing granulocytic sarcomas from lymphoid malignancies and also in the study of differentiation and transformation of human myeloid cells. The biological function of this antigen is not clear, although it has been proposed that it may play a role in the differentiation of myeloid cells.

Type: Primary
Antigen: Granulocyte Marker
Clonality: Monoclonal
Clone: BM-1
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR