Anti-PECAM1 Mouse Monoclonal Antibody (CF647) [clone: C31.7]

Supplier: Biotium
BNC470014-500 BNC470014-100
BNC470014-500EA 538 GBP
BNC470014-500 BNC470014-100
Anti-PECAM1 Mouse Monoclonal Antibody (CF647) [clone: C31.7]
Antibodies
CD31 (PECAM-1) is a transmembrane glycoprotein member of the immunoglobulin supergene family of adhesion molecules. CD31 is expressed by stem cells of the hematopoietic system and is primarily used to identify and concentrate these cells for experimental studies as well as for bone marrow transplantation. Anti-CD31 has shown to be highly specific and sensitive for vascular endothelial cells. Staining of nonvascular tumors (excluding hematopoietic neoplasms) is rare. CD31 MAb reacts with normal, benign, and malignant endothelial cells which make up blood vessel lining. The level of CD31 expression can help to determine the degree of tumor angiogenesis, and a high level of CD31 expression may imply a rapidly growing tumor and potentially a predictor of tumor recurrence.

Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: C31.7
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Cynomolgus monkey, Human, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR